1,2-Bis(dimethylsilyl)benzol

Lieferant: CHEMPUR SILANE

Synonyme: 1,2-Phenylenbis(dimethylsilan)

SILS01774.1 SILS01774.5 SILS01774.25
SILNSILS01774.1EA 0 EUR
SILNSILS01774.1 SILNSILS01774.5 SILNSILS01774.25
1,2-Bis(dimethylsilyl)benzol
1,2-Bis(dimethylsilyl)benzol
Formel: C₁₀H₁₈Si₂
Molecular Weight: 194.42 g/mol
MDL: MFCD00142462
CAS-Nummer: 17985-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR