1,4-Bis(dimethylsilyl)benzol
Lieferant: CHEMPUR SILANE
Synonyme:
1,4-Phenylenbis(dimethylsilan)
SILS01775.1
SILS01775.25
SILNSILS01775.1EA
0
EUR
SILNSILS01775.1
SILNSILS01775.25
1,4-Bis(dimethylsilyl)benzol
1,4-Bis(dimethylsilyl)benzol
Formel:
C₁₀H₁₈Si₂ Molecular Weight: 194.42 g/mol |
MDL:
MFCD00039790 CAS-Nummer: 2488-01-9 EINECS: 219-638-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...