N,N-Bis(trimethylsilyl)formamid

Lieferant: CHEMPUR SILANE

SILS02630.5 SILS02630.25
SILNSILS02630.5EA 0 EUR
SILNSILS02630.5 SILNSILS02630.25
N,N-Bis(trimethylsilyl)formamid
N,N-Bis(trimethylsilyl)formamid
Formel: C₇H₁₉NOSi₂
Molecular Weight: 189.4 g/mol
CAS-Nummer: 15500-60-4
EINECS: 239-530-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR