(3-Brompropyl)trichlorsilan
Lieferant: CHEMPUR SILANE
SILS03100.10
SILS03100.25
SILS03100.100
SILNSILS03100.10EA
0
EUR
SILNSILS03100.10
SILNSILS03100.25
SILNSILS03100.100
(3-Brompropyl)trichlorsilan
(3-Brompropyl)trichlorsilan
Formel:
BrH₂CCH₂CH₂SiCl₃ Molecular Weight: 256,43 g/mol Siedepunkt: 50 °C (0,001 mmHg) Dichte: 1,685 g/cm³ (20 °C) |
MDL:
MFCD00039291 CAS-Nummer: 13883-39-1 EINECS: 237-650-1 UN: 2987 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...