Butylsilan

Lieferant: CHEMPUR SILANE

SILS03485.10 SILS03485.25
SILNSILS03485.10EA 0 EUR
SILNSILS03485.10 SILNSILS03485.25
Butylsilan
Butylsilane
Formel: C₄H₁₂Si
Molecular Weight: 88,22 g/mol
MDL: MFCD00040079
CAS-Nummer: 1600-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR