(3-Chlorpropyl)trichlorsilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trichloro(3-chloropropyl)silane
SILS04625.25
SILS04625.100
SILNSILS04625.25EA
0
EUR
SILNSILS04625.25
SILNSILS04625.100
(3-Chlorpropyl)trichlorsilan
(3-Chlorpropyl)trichlorsilan
Formel:
ClH₂CCH₂CH₂SiCl₃ Molecular Weight: 211.98 g/mol Siedepunkt: 176…177 °C (1013 hPa) Dichte: 1.359 |
MDL:
MFCD00013694 CAS-Nummer: 2550-06-3 EINECS: 219-844-8 UN: 2987 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...