4-(Trichlorsilyl)butyronitril

Lieferant: CHEMPUR SILANE

Synonyme: Trichlor(3-cyanpropyl)silan, (3-Cyanpropyl)trichlorsilan

SILS05000.10 SILS05000.25 SILS05000.100
SILNSILS05000.10EA 0 EUR
SILNSILS05000.10 SILNSILS05000.25 SILNSILS05000.100
4-(Trichlorsilyl)butyronitril
4-(Trichlorsilyl)butyronitril
Formel: NCH₂CCH₂CH₂SiCl₃
Molecular Weight: 202,54 g/mol
Siedepunkt: 94 °C (1013 hPa)
Dichte: 1,28 g/cm³ (20 °C)
MDL: MFCD00013832
CAS-Nummer: 1071-27-8
EINECS: 213-990-6
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR