Cyclopentyltrichlorsilan

Lieferant: CHEMPUR SILANE

SILS05350.5 SILS05350.25
SILNSILS05350.5EA 0 EUR
SILNSILS05350.5 SILNSILS05350.25
Cyclopentyltrichlorsilan
Cyclopentyltrichlorsilan
Formel: C₅H₉Cl₃Si
Molecular Weight: 203.57 g/mol
Siedepunkt: 58 °C (1 mmHg)
Dichte: 1.2756 g/cm³ (20 °C)
MDL: MFCD00236508
CAS-Nummer: 14579-03-4
EINECS: 238-621-6
UN: 2987
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR