Decyltrichlorsilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trichloro(decyl)silane
SILS05575.10
SILS05575.25
SILS05575.100
SILNSILS05575.10EA
0
EUR
SILNSILS05575.10
SILNSILS05575.25
SILNSILS05575.100
Decyltrichlorsilan
Decyltrichlorsilan
Formel:
C₁₀H₂₁Cl₃Si Molecular Weight: 275.72 g/mol Siedepunkt: 183 °C (84 mmHg) Dichte: 1.054 g/cm³ (20 °C) |
MDL:
MFCD00013600 CAS-Nummer: 13829-21-5 EINECS: 237-540-3 UN: 2987 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...