N,N-Dimethyl-1,1,1-trimethylsilylamin

Lieferant: CHEMPUR SILANE

Synonyme: N-(Trimethylsilyl)dimethylamin, TMSDMA, N,N-Dimethyltrimethylsilylamin, N,N-Dimethylaminotrimethylsilan, Trimethylsilyldimethylamin, (Dimethylamino)trimethylsilan, Pentamethylsilylamin

SILS06925.10 SILS06925.25 SILS06925.100
SILNSILS06925.10EA 0 EUR
SILNSILS06925.10 SILNSILS06925.25 SILNSILS06925.100
N,N-Dimethyl-1,1,1-trimethylsilylamin
N,N-Dimethyl-1,1,1-trimethylsilylamin
Formel: C₅H₁₅NSi
Molecular Weight: 117,27 g/mol
Siedepunkt: 84 °C (1013 hPa)
Dichte: 0,7466 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008297
CAS-Nummer: 2083-91-2
EINECS: 218-222-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR