N,N-Diethyl-1,1-dimethylsilylamin

Lieferant: CHEMPUR SILANE

Synonyme: DMSDEA, N,N-Diethyldimethylsilylamine, N-(Dimethylsilyl)diethylamine, N,N-Diethylaminodimethylsilane, Dimethylsilyldiethylamine, (Diethylamino)dimethylsilane

SILS07500.5 SILS07500.25
SILNSILS07500.5EA 0 EUR
SILNSILS07500.5 SILNSILS07500.25
N,N-Diethyl-1,1-dimethylsilylamin
N,N-Diethyl-1,1-dimethylsilylamin
Formel: C₆H₁₇NSi
Molecular Weight: 131,29 g/mol
Siedepunkt: 109…110 °C (1013 hPa)
Dichte: 0,8 g/cm³ (20 °C)
MDL: MFCD00048523
CAS-Nummer: 13686-66-3
EINECS: 237-202-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR