Dodecyltrichlorsilan
Lieferant: CHEMPUR SILANE
SILS08525.25
SILS08525.100
SILNSILS08525.25EA
0
EUR
SILNSILS08525.25
SILNSILS08525.100
Dodecyltrichlorsilan
Dodecyltrichlorsilan
Formel:
C₁₂H₂₅Cl₃Si Siedepunkt: 293 °C (1013 hPa) Dichte: 1,03 g/cm³ (20 °C) |
MDL:
MFCD00000483 CAS-Nummer: 4484-72-4 EINECS: 224-769-9 UN: 1771 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...