Hexyltrichlorsilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trichlor(hexyl)silan, Hexylsilyltrichlorid
SILS09925.25
SILS09925.50
SILS09925.100
SILNSILS09925.25EA
0
EUR
SILNSILS09925.25
SILNSILS09925.50
SILNSILS09925.100
Hexyltrichlorsilan
Hexyltrichlorsilan
Formel:
C₆H₁₃Cl₃Si Molecular Weight: 219.61 g/mol Siedepunkt: 191.6 Dichte: 1.07 g/cm³ (20 °C) |
MDL:
MFCD00013606 CAS-Nummer: 928-65-4 EINECS: 213-178-1 UN: 1784 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...