Octyltrichlorsilan
Lieferant: CHEMPUR SILANE
SILS12750.25
SILS12750.100
SILNSILS12750.25EA
0
EUR
SILNSILS12750.25
SILNSILS12750.100
Octyltrichlorsilan
Octyltrichlorsilan
Formel:
C₈H₁₇Cl₃Si Siedepunkt: 233 °C (1013 hPa) Dichte: 1,07 g/cm³ (20 °C) |
MDL:
MFCD00000488 CAS-Nummer: 5283-66-9 EINECS: 226-112-1 UN: 1801 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...