Phenyltrimethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Trimethyl(phenyl)silan

SILS13775.25 SILS13775.100
SILNSILS13775.25EA 0 EUR
SILNSILS13775.25 SILNSILS13775.100
Phenyltrimethylsilan
Phenyltrimethylsilan
Formel: C₉H₁₄Si
Molecular Weight: 150,3 g/mol
Siedepunkt: 170 °C (1013 hPa)
Dichte: 0,873 g/cm³ (15 °C)
MDL: MFCD00008271
CAS-Nummer: 768-32-1
EINECS: 212-192-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR