Tetradecyltrichlorsilan

Lieferant: CHEMPUR SILANE

Synonyme: Trichloro(tetradecyl)silane

SILS15375.5 SILS15375.25 SILS15375.100
SILNSILS15375.5EA 0 EUR
SILNSILS15375.5 SILNSILS15375.25 SILNSILS15375.100
Tetradecyltrichlorsilan
Tetradecyltrichlorsilan
Formel: C₁₄H₂₉Cl₃Si
Molecular Weight: 331,83 g/mol
Siedepunkt: 131…132 °C (0,8 mmHg)
Dichte: 1 g/cm³ (20 °C)
MDL: MFCD00051440
CAS-Nummer: 18402-22-7
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR