(3,3,3-Trifluorpropyl)trichlorsilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trichlor(3,3,3-trifluorpropyl)silan
SILS17825.5
SILS17825.25
SILNSILS17825.5EA
0
EUR
SILNSILS17825.5
SILNSILS17825.25
(3,3,3-Trifluorpropyl)trichlorsilan
(3,3,3-Trifluorpropyl)trichlorsilan
Formel:
F₃CCH₂CH₂SiCl₃ Molecular Weight: 231,5 g/mol Siedepunkt: 111…112 °C (1013 hPa) Dichte: 1,4076 g/cm³ (20 °C) |
MDL:
MFCD00013602 CAS-Nummer: 592-09-6 EINECS: 209-744-2 UN: 2985 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...