Methoxytrimethylsilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trimethyl(methoxy)silan
SILS18650.10
SILS18650.25
SILNSILS18650.10EA
0
EUR
SILNSILS18650.10
SILNSILS18650.25
Methoxytrimethylsilan
Methoxytrimethylsilan
Formel:
C₄H₁₂OSi Molecular Weight: 104,22 g/mol Siedepunkt: 49…51 °C (1013 hPa) Dichte: 0,755 g/cm³ (20 °C) |
MDL:
MFCD00008344 CAS-Nummer: 1825-61-2 EINECS: 217-369-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...