2-(Trimethylsilyl)ethanol
Lieferant: CHEMPUR SILANE
SILS19375.5
SILS19375.25
SILNSILS19375.5EA
0
EUR
SILNSILS19375.5
SILNSILS19375.25
2-(Trimethylsilyl)ethanol
2-(Trimethylsilyl)ethanol
Formel:
HOH₂CCH₂Si(CH₃)₃ Molecular Weight: 118.25 g/mol Siedepunkt: 145 Schmelzpunkt: 0 Dichte: 0.8358 |
MDL:
MFCD00002825 CAS-Nummer: 2916-68-9 EINECS: 220-844-5 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...