Tri-tert-butylphosphin 10% (w/w) in Hexanen ≥99%

Lieferant: ABCR
Gefahr

Synonyme: Tris(1,1-dimethylethyl)phosphin

15-5811 15-5811-10G S15-5811.0010
ABCR15-5811EA 0 EUR
ABCR15-5811 STRE15-5811-10G ABCRS15-5811.0010
Tri-tert-butylphosphin 10% (w/w) in Hexanen ≥99%
Tri-tert-butylphosphin

Phosphine

Formel: C₁₂H₂₇P
Molecular Weight: 202,32 g/mol
Dichte: 0,680
MDL: MFCD00015006
CAS-Nummer: 13716-12-6
EINECS: 237-266-4
UN: 1993
ADR: 3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR