Anti-Mouse Monoclonal Antibody [clone: P3.6.2.8.1] (FITC)

Lieferant: Novus Biologicals
NOVUNBP1-43319FEA 628 EUR
NOVUNBP1-43319F
Anti-Mouse Monoclonal Antibody [clone: P3.6.2.8.1] (FITC)
Antikörper
Type: Control
Antigen:
Clonality: Monoclonal
Clone: P3.6.2.8.1
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR