N,N'-Ethylendianilin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,2-Dianilinoethan, N,N'-Diphenylethylendiamin
B23098.14
B23098.06
B23098.22
B23098.14EA
74.6
EUR
B23098.14
B23098.06
B23098.22
N,N'-Ethylendianilin ≥97%
N,N'-Ethylendianilin
Purity: 98+%
CAS No.: 150-61-8
Molecular Formula: C6H5NHCH2CH2NHC6H5
Formula Weight: 212.30
Melting Point: 64-68°
Boiling Point: 228°/12mm
Merck Reference: 14,2990
MDL No.: MFCD00003019
Formel:
C₆H₅NHCH₂CH₂NHC₆H₅ Molecular Weight: 212,29 g/mol Siedepunkt: 228 °C (12 mmHg) Schmelzpunkt: 64…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003019 CAS-Nummer: 150-61-8 EINECS: 205-765-6 Merck Index: 14,2990 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...