(4-Chlorphenyl)acetylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (4-Chlorphenyl)essigsäurechlorid

A12530.09 A12530.18 A12530.30
A12530.09EA 42.8 EUR
A12530.09 A12530.18 A12530.30
(4-Chlorphenyl)acetylchlorid ≥98%
(4-Chlorphenyl)acetylchlorid

Purity: 98% CAS No.: 25026-34-0 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Flash Point: 110°(230°F) Refractive Index: 1.5510 Boiling Point: 112-114°/11mm Beilstein Reference: 637775 UN No.: 3265 MDL No.: MFCD00037111

Formel: (C₆H₄Cl)CH₂COCl
Molecular Weight: 189,04 g/mol
Siedepunkt: 112…114 °C (11 mmHg)
Dichte: 1,292
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037111
CAS-Nummer: 25026-34-0
EINECS: 246-571-1
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR