(4-Fluorphenyl)acetylchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (4-Fluorphenyl)essigsäurechlorid

L19235.06 L19235.14
L19235.06EA 132 EUR
L19235.06 L19235.14
(4-Fluorphenyl)acetylchlorid ≥97%
(4-Fluorphenyl)acetylchlorid

Purity: 97% CAS No.: 459-04-1 Molecular Formula: C8H6ClFO Formula Weight: 172.59 Flash Point: 107°(224°F) Refractive Index: 1.5120 Boiling Point: 60°/8mm Beilstein Reference: 972148 UN No.: 3265 MDL No.: MFCD00674181

Formel: (C₆H₄F)CH₂COCl
Siedepunkt: 60 °C (8 mmHg)
Dichte: 1,259
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00674181
CAS-Nummer: 459-04-1
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR