(S)-(-)-β-Citronellol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-(-)-Citronellol
M-Clarity™ Qualitätsstufe = MQ100
303488-1G
303488-5G
SIAL303488-1GEA
58.1
EUR
SIAL303488-1G
SIAL303488-5G
(S)-(-)-β-Citronellol, Sigma-Aldrich®
(S)-(-)-β-Citronellol
Formel:
C₁₀H₂₀O Molecular Weight: 156,27 g/mol Dichte: 0,855 g/cm³ (20 °C) |
MDL:
MFCD00063214 CAS-Nummer: 7540-51-4 EINECS: 231-415-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...