1-(2-Nitrophenyl)piperidin

Lieferant: Thermo Scientific

BTB02207.1 BTB02207.10 BTB02207.50
MAYBBTB02207.1EA 0 EUR
MAYBBTB02207.1 MAYBBTB02207.10 MAYBBTB02207.50
1-(2-Nitrophenyl)piperidin
1-(2-Nitrophenyl)piperidin
Formel: C₁₁H₁₄N₂O₂
MDL: MFCD00023653
CAS-Nummer: 15822-77-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR