N,N'-Diethylethylendiamin 96%

Lieferant: Thermo Scientific
Gefahr

A17113.06 A17113.14 A17113.22
A17113.06EA 38.6 EUR
A17113.06 A17113.14 A17113.22
N,N'-Diethylethylendiamin 96%
N,N'-Diethylethylendiamin

Purity: 96% CAS No.: 111-74-0 Molecular Formula: CH3CH2NHCH2CH2NHCH2CH3 Formula Weight: 116.21 Flash Point: 33°(92°F) Refractive Index: 1.4326 Boiling Point: 152-154° UN No.: 2685 MDL No.: MFCD00009033

Formel: C₆H₁₆N₂
Molecular Weight: 116,21 g/mol
Siedepunkt: 152…154 °C
Schmelzpunkt: –11 °C
Dichte: 0,811
Flammpunkt: 33 °C (92 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009033
CAS-Nummer: 111-74-0
EINECS: 203-902-4
UN: 2685
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR