4,5-Dimethoxy-2-nitrobenzylalkohol ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
6-Nitroveratrumylalkohol, 6-Nitroveratrylalkohol, (4,5-Dimethoxy-2-nitrophenyl)methanol
L00719.06
L00719.14
L00719.06EA
88.2
EUR
L00719.06
L00719.14
4,5-Dimethoxy-2-nitrobenzylalkohol ≥98%
4,5-Dimethoxy-2-nitrobenzylalkohol
Purity: 98%
CAS No.: 1016-58-6
Molecular Formula: C9H11NO5
Formula Weight: 213.19
Storage Temperature: 2°C to 8°C
Melting Point: 144-148°
Beilstein Reference: 1880093
MDL No.: MFCD00014701
Formel:
C₉H₁₁NO₅ Molecular Weight: 213.19 Schmelzpunkt: 144…148 °C |
MDL:
MFCD00014701 CAS-Nummer: 1016-58-6 EINECS: 213-806-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...