3,5-Di-tert-butyl-4-hydroxybenzylalkohol ≥97%

Lieferant: Thermo Scientific
Achtung

A14709.06 A14709.14 A14709.22
A14709.06EA 50.3 EUR
A14709.06 A14709.14 A14709.22
3,5-Di-tert-butyl-4-hydroxybenzylalkohol ≥97%
3,5-Di-tert-butyl-4-hydroxybenzylalkohol

Purity: 97% CAS No.: 88-26-6 Molecular Formula: C15H24O2 Formula Weight: 236.36 Melting Point: 138-141° Boiling Point: 214°/40mm MDL No.: MFCD00017254

Formel: C₁₅H₂₄O₂
Molecular Weight: 236,35 g/mol
Siedepunkt: 214 °C (40 mmHg)
Schmelzpunkt: 138…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017254
CAS-Nummer: 88-26-6
EINECS: 201-815-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR