2,6-Bis(hydroxymethyl)-p-kresol
Lieferant: Thermo Scientific
Synonyme:
4-Methyl-2,6-bis(hydroxymethyl)phenol, 1,3-Bis(hydroxymethyl)-2-hydroxy-5-methylbenzene, 2,6-Bis(hydroxymethyl)-4-methylphenol, 3,5-Bis(hydroxymethyl)-4-hydroxytoluene
BTB10450.1
BTB10450.10
BTB10450.50
MAYBBTB10450.1EA
0
EUR
MAYBBTB10450.1
MAYBBTB10450.10
MAYBBTB10450.50
2,6-Bis(hydroxymethyl)-p-kresol
2,6-Bis(hydroxymethyl)-p-kresol
Formel:
C₉H₁₂O₃ Molecular Weight: 168.19 g/mol Siedepunkt: 363 Schmelzpunkt: 133…134 °C Dichte: 1.382 g/cm³ (–173 °C) |
MDL:
MFCD00004619 CAS-Nummer: 91-04-3 EINECS: 202-036-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...