2,6-Dichlor-4-nitrotoluol

Lieferant: Thermo Scientific

Synonyme: 1,3-Dichloro-2-methyl-5-nitrobenzene

KM08058DA KM08058EA
MAYBKM08058DAEA 0 EUR
MAYBKM08058DA MAYBKM08058EA
2,6-Dichlor-4-nitrotoluol
2,6-Dichlor-4-nitrotoluol
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,03 g/mol
MDL: MFCD00174267
CAS-Nummer: 7149-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR