4-Iod-2,5-xylenol

Lieferant: Thermo Scientific

Synonyme: 2,5-Dimethyl-4-iodphenol, 2-Hydroxy-5-iod-4-xylol, 5-Hydroxy-2-iod-4-xylol

RDP00233.1 RDP00233.10 RDP00233.50
MAYBRDP00233.1EA 0 EUR
MAYBRDP00233.1 MAYBRDP00233.10 MAYBRDP00233.50
4-Iod-2,5-xylenol
4-Iod-2,5-xylenol
Formel: C₈H₉IO
Molecular Weight: 248.06 g/mol
MDL: MFCD00219922
CAS-Nummer: 114971-53-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR