1,4-Phenyldiboronsäure

Lieferant: Thermo Scientific

Synonyme: 1,4-Benzoldiboronsäure, p-Phenylenbisboronsäure, Benzol-1,4-diboronsäure

AC35885.250 AC35885.1
MAYBAC35885.250EA 0 EUR
MAYBAC35885.250 MAYBAC35885.1
1,4-Phenyldiboronsäure
1,4-Phenyldiboronsäure
Formel: C₆H₈B₂O₄
Molecular Weight: 165,75 g/mol
MDL: MFCD00236018
CAS-Nummer: 4612-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR