16-Hydroxypalmitinsäure 97%
Lieferant: Thermo Scientific
Synonyme:
16-Hydroxyhexadecansäure
A17845.03
A17845.06
A17845.03EA
67.9
EUR
A17845.03
A17845.06
16-Hydroxypalmitinsäure 97%
16-Hydroxypalmitinsäure
Purity: 98%
CAS No.: 506-13-8
Molecular Formula: C16H32O3
Formula Weight: 272.43
Melting Point: 98-100°
Beilstein Reference: 1783998
Formel:
C₁₆H₃₂O₃ Molecular Weight: 272,43 g/mol Siedepunkt: 402 °C (1013 hPa) Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002750 CAS-Nummer: 506-13-8 EINECS: 208-028-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...