N,N-Diethylpropylendiamin 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-(Diethylamino)propylamin, DEAPA, N,N-Diethyl-1,3-diaminopropan, 3-Aminopropyldiethylamin, 1-Amino-3-(diethylamino)propan

L13888.AE L13888.AP
L13888.AEEA 23.5 EUR
L13888.AE L13888.AP
N,N-Diethylpropylendiamin 99%
N,N-Diethylpropylendiamin

Purity: 99% CAS No.: 104-78-9 Molecular Formula: C7H18N2 Formula Weight: 130.23 Flash Point: 51°(124°F) Refractive Index: 1.4420 Melting Point: -60° Boiling Point: 168-171° Beilstein Reference: 741879 UN No.: 2684 MDL No.: MFCD00008218

Formel: C₇H₁₈N₂
Molecular Weight: 130,23 g/mol
Siedepunkt: 168…171 °C
Schmelzpunkt: –60 °C
Dichte: 0,826
Flammpunkt: 51 °C (124 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008218
CAS-Nummer: 104-78-9
EINECS: 203-236-4
UN: 2684
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR