3-Brom-4-fluorbenzylalkohol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(3-Bromo-4-fluorophenyl)methanol
H30604.06
H30604.14
H30604.06EA
29.5
EUR
H30604.06
H30604.14
3-Brom-4-fluorbenzylalkohol ≥97%
3-Brom-4-fluorbenzylalkohol
Purity: 97%
CAS No.: 77771-03-0
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
Boiling Point: 80°/0.5mm
MDL No.: MFCD00143093
Formel:
C₇H₆BrFO Molecular Weight: 205,03 g/mol Siedepunkt: 80 °C (0,5 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143093 CAS-Nummer: 77771-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...