3-Brom-4-fluorbenzylalkohol

Lieferant: CHEMPUR

Synonyme: (3-Bromo-4-fluorophenyl)methanol

FL13217.POR 013217-25G 013217-5G
CHMPFL13217.POREA 0 EUR
CHMPFL13217.POR FLUO013217-25G FLUO013217-5G
3-Brom-4-fluorbenzylalkohol
3-Brom-4-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD00143093
CAS-Nummer: 77771-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR