1-Brom-2-pentin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

429538-10G 429538-1G
SIAL429538-10GEA 135 EUR
SIAL429538-10G SIAL429538-1G
1-Brom-2-pentin, Sigma-Aldrich®
1-Bromo-2-pentyne
Formel: C₅H₇Br
MDL: MFCD00236363
CAS-Nummer: 16400-32-1
EINECS: 240-451-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR