2-Brom-3,4-dimethoxybenzylalkohol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2-Brom-3,4-dimethoxyphenyl)methanol

342210050 342210250
ACRO342210050EA 24.9 EUR
ACRO342210050 ACRO342210250
2-Brom-3,4-dimethoxybenzylalkohol 98%
2-Brom-3,4-dimethoxybenzylalkohol
Formel: C₉H₁₁BrO₃
Molecular Weight: 247.09
Siedepunkt: 318
Schmelzpunkt: 95…100 °C
MDL: MFCD02093896
CAS-Nummer: 54370-00-2

Bestellen

Spezifikation Testergebnisse

Appearance White to pink-purple Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 100 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR