4-Brom-2-fluorbenzylalkohol

Lieferant: CHEMPUR

Synonyme: (4-Brom-2-fluorphenyl)methanol

FL13219.POR 013219-25G 013219-5G
CHMPFL13219.POREA 0 EUR
CHMPFL13219.POR FLUO013219-25G FLUO013219-5G
4-Brom-2-fluorbenzylalkohol
4-Brom-2-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD00143265
CAS-Nummer: 188582-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR