3-Brom-5-fluorbenzylalkohol
Lieferant: CHEMPUR
Synonyme:
(3-Bromo-5-fluorophenyl)methanol
FL13218.POR
013218-1G
013218-5G
CHMPFL13218.POREA
0
EUR
CHMPFL13218.POR
FLUO013218-1G
FLUO013218-5G
3-Brom-5-fluorbenzylalkohol
3-Brom-5-fluorbenzylalkohol
Formel:
C₇H₆BrFO Molecular Weight: 205,03 g/mol |
MDL:
MFCD04116320 CAS-Nummer: 216755-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...