2-Brom-5-fluorbenzylalkohol

Lieferant: CHEMPUR

Synonyme: (2-Bromo-5-fluorophenyl)methanol

FL13216.POR 013216-25G 013216-5G
CHMPFL13216.POREA 0 EUR
CHMPFL13216.POR FLUO013216-25G FLUO013216-5G
2-Brom-5-fluorbenzylalkohol
2-Brom-5-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD00142876
CAS-Nummer: 202865-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR