1-Butyl-3-methylimidazoliumdicyanamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-n-Butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyanamide

55220-50G-F 55220-5G-F
SIAL55220-50G-FEA 750 EUR
SIAL55220-50G-F SIAL55220-5G-F
1-Butyl-3-methylimidazoliumdicyanamid, Sigma-Aldrich®
1-Butyl-3-methylimidazolium dicyanamide
Formel: C₁₀H₁₅N₅
Molecular Weight: 205,26 g/mol
Dichte: 1,06 g/cm³ (20 °C)
MDL: MFCD07784457
CAS-Nummer: 448245-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR