2,2'-Methylenbis(4-chlorphenol) 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bis(5-chlor-2-hydroxyphenyl)methan, 2,2'-Methylenbis(4-chlorphenol)
A12640.22
A12640.14
A12640.36
A12640.22EA
49.4
EUR
A12640.22
A12640.14
A12640.36
2,2'-Methylenbis(4-chlorphenol) 95%
Dichlorophen
Purity: 95%
CAS No.: 97-23-4
Molecular Formula: C13H10Cl2O2
Formula Weight: 269.13
Melting Point: 172-176°
Beilstein Reference: 1884514
Merck Reference: 14,3071
UN No.: 3077
MDL No.: MFCD00002322
Formel:
C₁₃H₁₀Cl₂O₂ Siedepunkt: 389 °C (1013 hPa) Schmelzpunkt: 172…176 °C Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002322 CAS-Nummer: 97-23-4 EINECS: 202-567-1 ADR: 9,III Merck Index: 14,3071 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...