4-Chlor-2,6-xylenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A13649.09 A13649.18
A13649.09EA 46.4 EUR
A13649.09 A13649.18
4-Chlor-2,6-xylenol ≥98%
4-Chlor-2,6-xylenol

Purity: 98% CAS No.: 1123-63-3 Molecular Formula: C8H9ClO Formula Weight: 156.61 Melting Point: 80-83° Beilstein Reference: 1908282

Formel: C₈H₉ClO
Molecular Weight: 156.61 g/mol
Schmelzpunkt: 80…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020138
CAS-Nummer: 1123-63-3
EINECS: 214-376-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR