4-Chlor-2-methylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

647551-1G 647551-5G
SIAL647551-1GEA 56.2 EUR
SIAL647551-1G SIAL647551-5G
4-Chlor-2-methylbenzylalkohol, Sigma-Aldrich®
4-Chlor-2-methylbenzylalkohol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
MDL: MFCD06201140
CAS-Nummer: 129716-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR