4-Chlor-2-isopropyl-5-methylphenol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Chlorthymol
B21906.14
B21906.22
B21906.14EA
54.6
EUR
B21906.14
B21906.22
4-Chlor-2-isopropyl-5-methylphenol ≥99%
4-Chlor-2-isopropyl-5-methylphenol
Purity: 99%
CAS No.: 89-68-9
Molecular Formula: C10H13ClO
Formula Weight: 184.67
Melting Point: 59-61°
Boiling Point: 259-263°
Merck Reference: 14,2170
MDL No.: MFCD00002326
Formel:
C₁₀H₁₃ClO Molecular Weight: 184.67 g/mol Siedepunkt: 259…263 °C Schmelzpunkt: 59…61 °C Dichte: 1.228 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002326 CAS-Nummer: 89-68-9 EINECS: 201-930-1 Merck Index: 13,02189 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...