1-Ethyl-3-methylimidazoliumdicyanamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: EMIM DCA

M-Clarity™ Qualitätsstufe = MQ100

00796-50G-F 00796-5G-F
SIAL00796-50G-FEA 471 EUR
SIAL00796-50G-F SIAL00796-5G-F
1-Ethyl-3-methylimidazoliumdicyanamid, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium dicyanamide
Formel: C₈H₁₁N₅
Molecular Weight: 177.21 g/mol
MDL: MFCD08276373
CAS-Nummer: 370865-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR