3-Fluor-4-methylbenzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
(3-Fluoro-4-methylphenyl)methanol
005793-1G
005793-5G
FLUO005793-1GEA
0
EUR
FLUO005793-1G
FLUO005793-5G
3-Fluor-4-methylbenzylalkohol
3-Fluor-4-methylbenzylalkohol
Formel:
C₈H₉FO Molecular Weight: 140,16 g/mol |
MDL:
MFCD01631533 CAS-Nummer: 192702-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...