4-Fluor-3-methoxybenzylalkohol

Lieferant: CHEMPUR

Synonyme: (4-Fluoro-3-methoxyphenyl)methanol

FL13337.POR 013337-1G
CHMPFL13337.POREA 0 EUR
CHMPFL13337.POR FLUO013337-1G
4-Fluor-3-methoxybenzylalkohol
4-Fluor-3-methoxybenzylalkohol
Formel: C₈H₉FO₂
Molecular Weight: 156,16 g/mol
MDL: MFCD00272133
CAS-Nummer: 128495-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR